Learn More
Promega ALK2 Kinase Enzyme System, 1mg
Catalog No. PRV6272 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6272 1 mg
1 options
Catalog No. PRV6272 Supplier Promega Supplier No. V6272
Add to Cart
Add to Cart
Request a Quote
ALK2 is a receptor serine/threonine kinase that is member of the ALK family and is upstream of signaling pathway involving the SMAD proteins especially SMAD1/5/8.
  • Easily Screen and Profile ALK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C