Learn More
Promega ALK4 Kinase Enzyme System, 10microg
Catalog No. PRV4508 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4508 10 μg
1 options
Catalog No. PRV4508 Supplier Promega Supplier No. V4508
Recombinant human ALK4 (150-end) was expressed in Sf9 cells using an N-terminal GST tag. ALK4 is a member of the subfamily of receptor ser/thr kinases that mediates signaling by the Activins.
  • Easily Screen and Profile ALK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C