Learn More
Promega ALK4 Kinase Enzyme System, 1mg
Catalog No. PRV6575 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6575 1 mg
1 options
Catalog No. PRV6575 Supplier Promega Supplier No. V6575
Add to Cart
Add to Cart
Request a Quote
Recombinant human ALK4 (150-end) was expressed in Sf9 cells using an N-terminal GST tag. ALK4 is a member of the subfamily of receptor ser/thr kinases that mediates signaling by the Activins.
  • Easily Screen and Profile ALK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C