Learn More
Promega ALK6 Kinase Enzyme System, 1mg
Catalog No. PRV6273 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6273 1 mg
1 options
Catalog No. PRV6273 Supplier Promega Supplier No. V6273
Add to Cart
Add to Cart
Request a Quote
ALK6 (also known as BMPR1B) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2.
  • Easily Screen and Profile ALK6 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C