Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline-Iodide Reagent, Winkler Formulation, Ricca Chemical

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Specifications
Specifications
Color | Clear colorless |
pH Range | >13 |
Volume (Metric) | 100 mL |
Packaging | Natural Poly Bottle |
Quantity | 100 mL |
Type | Alkaline-Iodide Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction