Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase/ALPL Antibody (071), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289224JF646
Description
Alkaline Phosphatase/ALPL Monoclonal antibody specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Western BlotSpecifications
| Alkaline Phosphatase/ALPL | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 071 | |
| Western Blot | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Alkaline Phosphatase/ALPL. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 249 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction