Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (8B6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 11 publications
$192.94 - $530.25
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | 8B6 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry reported in multiple pieces of scientific literature, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1103638SS
![]() |
Novus Biologicals
NB1103638SS |
0.025 mL |
Each for $192.94
|
|
|||||
NB1103638
![]() |
Novus Biologicals
NB1103638 |
0.1 mL |
Each for $530.25
|
|
|||||
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunodiffusion.Specifications
| Alkaline Phosphatase/ALPP | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| P05187 | |
| 250 | |
| Hep-2 cells with boosted surface expression of Alkaline Phosphatase, Placental. [UniProt# P05187]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 70 kDa |
| 8B6 | |
| Western Blot 1:1000, Flow Cytometry reported in multiple pieces of scientific literature, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunodiffusion | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| ALPP | |
| IgG2a κ | |
| Protein A purified | |
| Alkaline Phosphatase, Placental - both Regan and Nagao isoenzymes. No cross reactivity with other isoenzymes of Alkaline Phosphatase. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title