Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (ALPP/516), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $487.50
Specifications
Antigen | Alkaline Phosphatase/ALPP |
---|---|
Clone | ALPP/516 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 1.0 - 2.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24495700
![]() |
Novus Biologicals
NBP2449570.02MG |
0.02 mg |
Each for $243.50
|
|
|||||
NBP24495701
![]() |
Novus Biologicals
NBP2449570.1MG |
0.1 mg |
Each for $487.50
|
|
|||||
NBP24495702
![]() |
Novus Biologicals
NBP2449570.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Frozen), Immunofluorescence
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA.Specifications
Alkaline Phosphatase/ALPP | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
ALPP | |
IgG1 κ | |
Protein A or G purified | |
PLAP is a tissue specific, trophoblast-derived, 70kDa, glycosyl-phosphatidylinositol (GPI)-anchored, dimeric, Zn2+ metallo-glycoprotein that catalyzes the hydrolysis of phosphomonoesters into an inorganic phosphate and an alcohol. It is present in the placenta and serum of pregnant women and in high frequency in gynecological and testicular cancers and in lower frequency in other tumors. The three tissue-specific AP's in humans, PLAP, germ cell AP (GCAP) and intestinal AP, are 90-98% homologous. Non-tissue specific AP is found in kidney, liver and bone. This MAb binds equally well to all common allelic variants (S, F, FS and I) of PLAP and to some variants of AP from normal human testis. This MAb can be used as tracer antibody in ELISA to detect PLAP in serum of S, F, FS and I phenotypes. |
ALPP/516 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Frozen 1.0 - 2.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P05187 | |
250 | |
Recombinant full-length human ALPP protein | |
Primary | |
Store at 4C. | |
70 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title