Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (HD 11F7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$172.57 - $418.55
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | HD 11F7 |
| Concentration | 1 mg/mL |
| Dilution | ELISA, Immunohistochemistry : 1:10 - 1:500, Radioimmunoassay : 1:100 - 1:2000 |
| Applications | ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Immunohistochemistry (Frozen), Radioimmune Assays (RIA) |
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay.Specifications
| Alkaline Phosphatase/ALPP | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Immunohistochemistry (Frozen), Radioimmune Assays (RIA) | |
| Unconjugated | |
| Human | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| ALPP | |
| IgG2b | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| HD 11F7 | |
| ELISA, Immunohistochemistry : 1:10 - 1:500, Radioimmunoassay : 1:100 - 1:2000 | |
| Monoclonal | |
| Mouse | |
| PBS with 0.02% sodium azide | |
| 250 | |
| Fresh Hep-2 cells followed by booster injections of fresh Hep-2 cells that had their surface expression of Alkaline Phosphatase/ALPP boosted in culture. | |
| Primary | |
| This antibody recognizes both Regan and Nagao isoenzymes of placental alkaline phosphatase (PLAP). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title