| Antigen |
Alkaline Phosphatase/ALPP |
| Applications |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification |
Polyclonal |
| Concentration |
1 mg/ml |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml |
| Formulation |
PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. |
P05187 |
| Gene Alias |
Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme |
| Gene Symbols |
ALPP |
| Host Species |
Rabbit |
| Immunogen |
Synthetic peptides corresponding to ALPP(alkaline phosphatase, placental (Regan isozyme)) The peptide sequence was selected from the C terminal of ALPP (NP_001623). Peptide sequence TVLLYGNGPGYVLKDGARPDVTESESGSPEYRQQSAVPLDEETHAGEDVA. |
| Molecular Weight of Antigen |
59 kDa |
| Purification Method |
Protein A purified |
| Quantity |
100 μL |
| Regulatory Status |
RUO |
| Research Discipline |
Embryonic Stem Cell Markers, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
250 |
| Test Specificity |
Expected identity based on immunogen sequence: Human: 100%; Xenopus: 85%; Canine: 85%; Western clawed frog: 85%; Green puffer: 85%; Yellowfever mosquito: 84%; Shewanella baltica BA175: 84%; Shewanella baltica OS183: 84%; Shewanella baltica OS678: 84%; Shewanella benthica KT99: 84%; Pseudoalteromonas tunicata D2: 84%; Alteromonadales bacterium TW-7: 84%; Sea squirt: 78%; Chicken: 78%; Zebrafish: 78%; Mouse: 78%; Atlantic cod: 78%; Rat: 78%; Silk moth: 78%; Wild silk moth: 78%; Bovine: 78%; Florida lancelet: 78%; Savannah tsetse fly: 78%; Congregibacter litoralis KT71: 76%; Black-legged tick: 76%; Red flour beetle: 76%; Gamma proteobacterium HTCC2207: 76%; Fruit fly: 76%; Encephalitis mosquito: 75%;. |
| Reconstitution |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. |
| Target Species |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish |
| Content And Storage |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Form |
Purified |
| Isotype |
IgG |
|
Show More
Show Less
|
|