Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase/ALPP Rabbit anti-Human, Clone: ALPP/2899R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | ALPP/2899R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Alkaline Phosphatase/ALPP | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 250 | |
| IgG | |
| Protein A or G purified |
| ALPP/2899R | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 0.25-0.5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| Recombinant full-length human Alkaline Phosphatase/ALPP protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title