Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AAP1), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002638G
Description
Alkaline Phosphatase, Intestinal Monoclonal specifically detects Alkaline Phosphatase, Intestinal in Human samples. It is validated for ELISA.Specifications
Alkaline Phosphatase, Intestinal | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ALPI | |
D98/AH-2 (HeLa) cells, which produce human intestinal alkaline phosphatase ectopically | |
0.1 mL | |
Cancer, Embryonic Stem Cell Markers, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers | |
248 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA | |
AAP1 | |
Western Blot, ELISA | |
alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction