Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AP-7/6/7), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250102PCP
Description
Alkaline Phosphatase, Intestinal Monoclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Alkaline Phosphatase, Intestinal | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
| Immunohistochemistry, Immunocytochemistry | |
| AP-7/6/7 | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| Calf intestinal alkaline phosphatase | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Ovarian Carcinoma Cell Markers, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 248 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction