Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (SR2122), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321913100UL
Description
Alkaline Phosphatase, Intestinal Monoclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Alkaline Phosphatase, Intestinal | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR2122 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
A synthesized peptide derived from human Alkaline Phosphatase, Intestinal (Uniprot #: P09923) | |
100 μg | |
Cancer, Lipid and Metabolism, Ovarian Carcinoma Cell Markers, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
248 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction