Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Alkaline Phosphatase (Isoform 4) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585878
Description
Alkaline Phosphatase (Isoform 4) Polyclonal Antibody for Western Blot
Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism's environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.
Specifications
Alkaline Phosphatase (Isoform 4) | |
Polyclonal | |
Unconjugated | |
ALPP | |
alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; ALPP; glycerophosphatase; PALP; Placental alkaline phosphatase 1; PLAP; PLAP-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P05187 | |
ALPP | |
Synthetic peptide within Human Alkaline Phosphatase aa 314-363. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction