Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066385
Description
Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Human, Bacteria samples. It is validated for Western Blot, ELISA.Specifications
| Alkaline Phosphatase, Liver/Bone/Kidney | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| ALPL, BALP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 249 | |
| Human, Bacteria | |
| Purified |
| Western Blot, ELISA | |
| BGN/03/662 | |
| Unconjugated | |
| P05186 | |
| ALPL | |
| Alkaline phosphatase from human bone. | |
| 0.1 mg | |
| Primary | |
| NB100-66385 recognizes bone specific alkaline phosphatase. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction