Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066385F
Description
Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Human, Bacteria samples. It is validated for ELISA.Specifications
Alkaline Phosphatase, Liver/Bone/Kidney | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ALPL | |
Alkaline phosphatase from human bone. | |
0.1 mL | |
Primary | |
NB100-66385 recognizes bone specific alkaline phosphatase. | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
BGN/03/662 | |
ELISA | |
ALPL, BALP | |
Mouse | |
Protein A purified | |
RUO | |
249 | |
Human, Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction