Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alkaline Phosphatase, Tissue Non-Specific Mouse anti-Bovine, Human, Clone-ALPL/597, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
| Antigen | Alkaline Phosphatase, Tissue Non-Specific |
|---|---|
| Clone | ALPL/597 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:100 - 1:200, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254376100
![]() |
Novus Biologicals
NBP254376100UG |
100 μg |
Each for $646.00
|
|
|||||
NB254376200
![]() |
Novus Biologicals
NBP254376200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Alkaline Phosphatase, Tissue Non-Specific Monoclonal specifically detects Alkaline Phosphatase, Tissue Non-Specific in Human, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Alkaline Phosphatase, Tissue Non-Specific | |
| 1 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Bovine | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| ALPL | |
| IgG1 κ | |
| Protein A or G purified | |
| 55 kDa |
| ALPL/597 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:100 - 1:200, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Protein Phosphatase | |
| PBS with No Preservative | |
| 249 | |
| Recombinant human ALPL protein | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title