Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565405 Supplier Invitrogen™ Supplier No. PA565405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDERELAPGSP IASVSFGACR DFVFRHKDSR GKSPSRRVAV VRLPLAHGSL LMMNHPTNTH WYHSLPVRKK VLAPRVNLTF RK Highest antigen sequence identity to the following orthologs - mouse 89%, rat 88%.

The E. coli AlkB protein protects against the cytotoxicity of methylating agents by repair of the specific DNA lesions generated in single-stranded DNA; ALKBH2 and ALKBH3 are mammalian homologs of AlkB that catalyze the removal of 1-methyladenine and 3-methylcytosine, modifications that left unchecked could lead to cancerous cells. Mutations in both ALKBH2 and ALKBH3 have been observed in pediatric brain tumors indicating that these proteins are important in the prevention of cancer formation. Like the histone demethylase JMJD1A, ALKBH2 is a non-heme iron enzyme that is inhibited by Nickel ions, suggesting that inhibition of ALKBH2 by Nickel ions may play a role in the development of cancer. Conversely, ALKBH2 mRNA and protein levels are increased glioma cells following Photofrin-mediated photodynamic therapy, an adjuvant therapy in cancer treatment, suggesting that down-regulating ALKBH2 expression in cancer cells may enhance the anti-cancer effectiveness of this treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALKBH2
Gene Accession No. Q6NS38
Gene Alias 2OG-Fe(II) oxy DC1; 9530023G02; Abh2; AlkB homolog 2; alkB homolog 2, alpha-ketoglutarate dependent dioxygenase; alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 2; alkB, alkylation repair homolog 2 (E. coli); ALKBH2; Alkylated DNA repair protein alkB homolog 2; alpha-ketoglutarate-dependent dioxygenase alkB homolog 2; AU016977; DNA oxidative demethylase ALKBH2; mABH2; oxy DC1; RGD1306377
Gene Symbols ALKBH2
Host Species Rabbit
Immunogen Recombinant Human ALKBH2. Recombinant protein control fragment (Product #RP-106087).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121642
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.