Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALKBH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521005
Description
A suggested positive control is human kidney tissue lysate. PA5-21005 can be used with blocking peptide PEP-1119.
The E. coli AlkB protein protects against the cytotoxicity of methylating agents by repair of the specific DNA lesions generated in single-stranded DNA; ALKBH2 and ALKBH3 are mammalian homologs of AlkB that catalyze the removal of 1-methyladenine and 3-methylcytosine, modifications that left unchecked could lead to cancerous cells. Mutations in both ALKBH2 and ALKBH3 have been observed in pediatric brain tumors indicating that these proteins are important in the prevention of cancer formation. Like the histone demethylase JMJD1A, ALKBH2 is a non-heme iron enzyme that is inhibited by Nickel ions, suggesting that inhibition of ALKBH2 by Nickel ions may play a role in the development of cancer. Conversely, ALKBH2 mRNA and protein levels are increased glioma cells following Photofrin-mediated photodynamic therapy, an adjuvant therapy in cancer treatment, suggesting that down-regulating ALKBH2 expression in cancer cells may enhance the anti-cancer effectiveness of this treatment.
Specifications
ALKBH2 | |
Polyclonal | |
Unconjugated | |
ALKBH2 | |
2OG-Fe(II) oxy DC1; 9530023G02; Abh2; AlkB homolog 2; alkB homolog 2, alpha-ketoglutarate dependent dioxygenase; alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 2; alkB, alkylation repair homolog 2 (E. coli); ALKBH2; Alkylated DNA repair protein alkB homolog 2; alpha-ketoglutarate-dependent dioxygenase alkB homolog 2; AU016977; DNA oxidative demethylase ALKBH2; mABH2; oxy DC1; RGD1306377 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
121642, 231642, 304578 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6NS38, Q6P6J4 | |
ALKBH2 | |
A 15 amino acid peptide near the carboxy terminus of human ALKBH2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction