Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100786 Supplier Invitrogen™ Supplier No. PA5100786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ALKBH3.

Dioxygenase that repairs alkylated DNA containing 1- methyladenine (1meA) and 3-methylcytosine (3meC) by oxidative demethylation. Has a strong preference for single-stranded DNA. Able to process alkylated 3mC within double-stranded regions via its interaction with ASCC3, which promotes DNA unwinding to generate single-stranded substrate needed for ALKHB3. May also act on RNA. Requires molecular oxygen, alpha-ketoglutarate and iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Alkbh3
Gene Accession No. Q5XIC8, Q8K1E6, Q96Q83
Gene Alias 1700108H04Rik; 1810020C19Rik; Abh3; AlkB homolog 3; alkB homolog 3, alpha-ketoglutaratedependent dioxygenase; alkB homolog 3, alpha-ketoglutarate-dependent dioxygenase; alkB, alkylation repair homolog 3; alkB, alkylation repair homolog 3 (E. coli); Alkbh3; Alkylated DNA repair protein alkB homolog 3; alpha-ketoglutarate-dependent dioxygenase alkB homolog 3; DEPC 1; DEPC1; DEPC-1; FLJ43614; hABH3; m MGC118790; mABH3; MGC118792; MGC118793; MGC134125; PCA1; Prostate cancer antigen 1; prostate cancer antigen-1; RP23-375N21.4
Gene Symbols Alkbh3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALKBH3(Accession Q96Q83), corresponding to amino acid residues R164-T214.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221120, 362169, 69113
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.