Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562407 Supplier Invitrogen™ Supplier No. PA562407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYRFIYCSDT GWAVGTEESD FEGWAFPFPG VMLIEDFVTR EEEAELVRLM DRDPWKLSQS GRRKQDYGPK VN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

Dioxygenase that mediates demethylation of actin monomethylated at 'Lys-84' (K84me1), thereby acting as a regulator of actomyosin-processes (PubMed:23673617). Demethylation of actin K84me1 is required for maintaining actomyosin dynamics supporting normal cleavage furrow ingression during cytokinesis and cell migration (PubMed:23673617). In addition to proteins, also demethylates DNA: specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA, thereby regulating Polycomb silencing. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALKBH4
Gene Accession No. Q9NXW9
Gene Alias 2010004B12Rik; ABH4; Abj4; alkB homolog 4, lysine demethylase; alkB homolog 4, lysine demthylase; alkB, alkylation repair homolog 4; alkB, alkylation repair homolog 4 (E. coli); Alkbh4; Alkylated DNA repair protein alkB homolog 4; alpha-ketoglutarate-dependent dioxygenase alkB homolog 4; DNA N6-methyl adenine demethylase ALKBH4; Lysine-specific demethylase ALKBH4; probable alpha-ketoglutarate-dependent dioxygenase ABH4; RGD1308608
Gene Symbols ALKBH4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALKBH4. Recombinant protein control fragment (Product #RP-98508).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54784
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.