Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALKBH5 Mouse anti-Human, Mouse, Rat, Clone: 2D2B2, ProteinTech
SDP

Catalog No. 502239860 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9860 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9860 Supplier Proteintech Group Inc Supplier No. 678111IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aliquoting is unnecessary for -20°C storage.

ALKBH5 Monoclonal antibody specifically detects ALKBH5 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen ALKBH5
Applications Western Blot
Classification Monoclonal
Clone 2D2B2
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ALKBH5
Gene Accession No. D3ZKD3, Q3TSG4, Q6P6C2
Gene Alias ABH5; AlkB family member 5, RNA demethylase; alkB homolog 5, RNA demethylase; alkB, alkylation repair homolog 5; alkB, alkylation repair homolog 5 (E. coli); Alkbh5; Alkylated DNA repair protein alkB homolog 5; alpha-ketoglutarate-dependent dioxygenase alkB homolog 5; AW050020; AW215868; E130207K11; OFOXD; OFOXD1; oxoglutarate and iron-dependent oxygenase domain containing; probable alpha-ketoglutarate-dependent dioxygenase ABH5; RGD1309496; RNA demethylase ALKBH5
Gene Symbols ALKBH5
Host Species Mouse
Immunogen ALKBH5 Fusion Protein Ag10490
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268420, 303193, 54890
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.