Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588210 Supplier Invitrogen™ Supplier No. PA588210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, LO2, HeLa; Cellular Location: Nucleus speckle Immunogen sequence: SLPVRRGSVT VLSGYAADEI THCIRPQDIK ERRAVIILRK TRLDAPRLET KSLSSSVLPP SYASDRLSGN NRDPALKPKR SHRKADPDAA HRPRILEMDK EENRRSVLLP THRRRGSFSS ENYWRKSYES SEDCSEAAGS PARKVKMRRH.

ALKBH5 (RNA demehtylase ALKBH5) is a dioxygenase that demethylates RNA by oxidative demethylation. Specifically, it demethylates N(6)-methyladenosine (m6A) RNA, the most prevalent internal modification of messenger RNA (mRNA) in higher eukaryotes. Demethylation of m6A mRNA affects mRNA processing and export. ALKBH5 can also demethylate N(6)-methyladenosine in single-stranded DNA (in vitro). It requires molecular oxygen, alpha-ketoglutarate and iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALKBH5
Gene Accession No. D3ZKD3, Q3TSG4, Q6P6C2
Gene Alias ABH5; AlkB family member 5, RNA demethylase; alkB homolog 5, RNA demethylase; alkB, alkylation repair homolog 5; alkB, alkylation repair homolog 5 (E. coli); Alkbh5; Alkylated DNA repair protein alkB homolog 5; alpha-ketoglutarate-dependent dioxygenase alkB homolog 5; AW050020; AW215868; E130207K11; OFOXD; OFOXD1; oxoglutarate and iron-dependent oxygenase domain containing; probable alpha-ketoglutarate-dependent dioxygenase ABH5; RGD1309496; RNA demethylase ALKBH5
Gene Symbols ALKBH5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 245-394 of human ALKBH5 (NP_0602283).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268420, 303193, 54890
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.