Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ALKBH5, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1013IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1013IMI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1013IMI Supplier MilliporeSigma™ Supplier No. ABE1013I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA demethylase ALKBH5 (UniProt Q6P6C2; also known as Alkylated DNA repair protein alkB homolog 5, Alpha-ketoglutarate-dependent dioxygenase alkB homolog 5) is encoded by the ALKBH5 (also known as ABH5, OFOXD1) gene (Gene ID 54890) in human. More than 100 types of postranscriptional RNA modifications are known, among which N6-methyladenosine (m6A) is the most common RNA modification. m6A modification mainly occurs in the consensus seqeunce motif RRm6ACH (R = A/G and H = A/C/U) located in stop codons, long internal exons as well as 3&prime UTR. RNA methyltransferases and demethylases are responsible for maintaining the level of cellular RNA m6A methylation. Two Fe2+ and 2-oxoglutarate (2OG) dependent AlkB dioxygenases family members, the fat mass and obesity-associated (FTO) enzyme and ALKBH5, catalyze the oxidative demethylation of abundant m6A residues. Both FTO and ALKBH5 are localized to nucleus and ALKBH5 gene knockdown is shown to affect cellular mRNA splicing in vitro and result in sterility phenotype in male mice in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH5
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ALKBH5, ABH5, OFOXD1
Gene Accession No. Q6P6C2
Host Species Rabbit
Immunogen GST-tagged recombinant human ALKBH5 C-terminal fragment.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060228
Test Specificity Expected to react with all three spliced isoforms of human ALKBH5 reported by UniProt (Q6P6C2).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.