Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALKBH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567339 Supplier Invitrogen™ Supplier No. PA567339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NWGGLPHPRGM VPERLPPWLQ RYVDKVSNLS LFGGLPANHV LVNQYLPGEG IMPHEDGPLY YPTVSTISLG SHTVLDFYEP RRPEDDDPTE Q Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

Probable dioxygenase that requires molecular oxygen, alpha-ketoglutarate and iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALKBH6
Gene Accession No. Q3KRA9
Gene Alias ABH6; AI839550; alkB homolog 6; alkB, alkylation repair homolog 6; alkB, alkylation repair homolog 6 (E. coli); ALKBH6; Alkylated DNA repair protein alkB homolog 6; alkylation repair homolog 6; alpha-ketoglutarate-dependent dioxygenase alkB homolog 6; C130099A02; probable alpha-ketoglutarate-dependent dioxygenase ABH6
Gene Symbols ALKBH6
Host Species Rabbit
Immunogen Recombinant Human ALKBH6. Recombinant protein control fragment (Product #RP-107988).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84964
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.