Learn More
Invitrogen™ ALKBH6 Polyclonal Antibody

Description
Immunogen sequence: GMGMLNLEIGG DAGGRIGCKE LVLMEEQDAR VPALEPFRVE QAPPVIYYVP DFISKEEEEY LLRQVFNAPK PKWTQLS Highest antigen sequence identity to the following orthologs - mouse 68%, rat 67%.
Specifications
Specifications
| Antigen | ALKBH6 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ALKBH6 |
| Gene Accession No. | Q3KRA9 |
| Gene Alias | ABH6; AI839550; alkB homolog 6; alkB, alkylation repair homolog 6; alkB, alkylation repair homolog 6 (E. coli); ALKBH6; Alkylated DNA repair protein alkB homolog 6; alkylation repair homolog 6; alpha-ketoglutarate-dependent dioxygenase alkB homolog 6; C130099A02; probable alpha-ketoglutarate-dependent dioxygenase ABH6 |
| Gene Symbols | ALKBH6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.