Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALKBH6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571509
Description
Probable dioxygenase that requires molecular oxygen, alpha-ketoglutarate and iron (By similarity).
Specifications
ALKBH6 | |
Polyclonal | |
Unconjugated | |
ALKBH6 | |
Alpha-ketoglutarate-dependent dioxygenase alkB homolog 6; 11411-; Alkylated DNA repair protein alkB homolog 6; ALKBH6; ABH6 | |
Rabbit | |
Protein A | |
RUO | |
233065, 84964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q3KRA9, Q8K2U2 | |
ALKBH6 | |
KLH conjugated synthetic peptide between 116-145 amino acids from the Central region of human ALKBH6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction