Learn More
United Scientific™ All Glass Vacuum Filter Holder Set and Accessories
Shop All United Scientific Supplies, Inc Products
Description
- Used in particulate contamination analysis for filtering aqueous, organic, or corrosive liquids.
- Recommended for HPLC solvent filtration.
- Complete set includes a borosilicate glass funnel, anodized aluminum spring clamp, vacuum base with sintered disc, and ground joint flask.
- Membrane filters are not included.
- Parts are also available separately.
Specifications
Specifications
| For Use With (Application) | Particulate Contamination Analysis |
| Includes | Borosilicate Glass Funnel, Anodized Aluminum Spring Clamp, Vacuum Base with Sintered Disc, and Ground Joint Flask |
| Item Description | Glass Vacuum Filter Holder |
| Material | Borosilicate Glass |
| Product Type | Complete Filter Holder Set |
| Standard Taper Joint No. | 34/35 |