Learn More
Cole-Parmer™ All-in-One pH/ATC Electrode, Double-Junction
Catalog No. 50633526 Shop All Oakton Products
Change view
Click to view available options
For Use With (Equipment):
Oakton Waterproof 600 Series Meters
Catalog No. For Use With (Equipment)
50633526 Oakton Waterproof 600 Series Meters
1 options
Catalog No. 50633526 Supplier Cole-Parmer™ Supplier No. 3581672
Add to Cart
Add to Cart
Replace your All-in-One pH/ATC electrode when it breaks, no longer holds calibration, cannot be rehydrated, or is not compatible with your sample.
  • Epoxy body
  • Shielded bulb for durability
  • Gel-filled
  • Double-junction for dirty samples
Specifications
Type Electrode, pH/ATC Combination
Description All-in-One pH/ATC Electrode for 600 series meters, Double-Junction
Interface BNC / 8-Pin Waterproof
Material Epoxy
Warranty 1 Year
For Use With (Application) Dirty samples and rugged fieldwork
For Use With (Equipment) Oakton Waterproof 600 Series Meters
Range 0 to 12 pH
Temperature Range (English) 32°F to 176°F
Temperature Range (Metric) 0°C to 80°C
Length (English) Cable 30 in.