Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Westcott™ All Purpose Value Scissors
SCI_ED

Catalog No. p-7214865
Click to view available options
Description:
Bent
Straight

Scissors with stainless steel blades

  • Stainless steel blades
  • Bent handle cuts easily and comfortably on a flat surface
  • Perfect for light use, cutting through paper, ribbon and photos
  • Limited Five-Year Manufacturers Warranty

Specifications

Product Type Scissor
Color Black
Description Bent
Grade Pre-K to 12
Age 4 to 18
Quantity 3/Pk.
Length (English) 8 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.