Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals all-trans-Retinol, 95%

Catalog No. AC347071000
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 68-26-8 | C20H30O | 286.459 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications


• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-26-8
Molecular Formula C20H30O
Molecular Weight (g/mol) 286.459
MDL Number MFCD00001552
InChI Key FPIPGXGPPPQFEQ-OVSJKPMPSA-N
Synonym retinol, vitamin a, all-trans-retinol, vitamin a1, alphalin, axerophthol, vitamin a alcohol, oleovitamin a, chocola a, alphasterol
PubChem CID 445354
ChEBI CHEBI:17336
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraen-1-ol
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCO)C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 mg
Beilstein 06, III, 2787
Merck Index 15, 10211
Formula Weight 286.46
Percent Purity ≥94%
Physical Form Powder or Lumps
Chemical Name or Material all-trans-Retinol, 95%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-683-7
RTECSNumber VH6750000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.