Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized

Catalog No. ac346321000
Click to view available options
Quantity:
100 g

Catalog No. AC346321000

Supplier: Thermo Scientific Chemicals 346321000

Only null left
Add to Cart

CAS: 127-47-9 | C22H32O2 | 328.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-47-9
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.49
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
Synonym retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
PubChem CID 638034
ChEBI CHEBI:32095
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Flash Point >100°C
Packaging Glass bottle
Refractive Index 1.5470 to 1.5550
Quantity 100 g
Merck Index 15,10211
Solubility Information Solubility in water: difficult soluble
Viscosity 20 mPa.s (60°C)
Formula Weight 328.49
Related Substances 0.14 (A370/A326) max
Chemical Name or Material all-trans-Retinyl acetate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 204-844-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.