Learn More
Allegro™ 9872 FIlters
Description
- Replacement filter kit 9872‐50
- For use with 2 and 5-Worker
- Portable or wall-mount 9872 filtration panels
- Filter kit includes: Two 1st stage, one 2nd stage and one 3rd stage filter
Specifications
Specifications
| For Use With (Equipment) | Carry-air and Wall-air Model 9872 Filtration Panels |
| Includes | Two 1st stage, one 2nd stage and one 3rd stage filter |
| Model | 987250 |
| Product Type | Filter Kit |