Learn More
Allegro™ Suspenders for Back Supports
Specifications
Specifications
| Color | Black |
| Closure Type | Hook and loop |
| Product Type | Suspender |
| Size | Large |
| For Use With (Application) | Designed to be used with back support which helps to reduce back stress and strain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.