Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allergin-1 Antibody (767727), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7638MFV500
Description
Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| Allergin-1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284021 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 767727 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog | |
| Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227, Accession # Q7Z6M3 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction