Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allergin-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Allergin-1 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Allergin-1 Polyclonal specifically detects Allergin-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Allergin-1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Human | |
Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog | |
MILR1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
284021.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RKAVLDCEAMKTNEFPSPCLDSKTKVVMKGQNVSMFCSHKNKSLQITYSLFRRKTHLGTQDGKGEPAIFNLSITEAHESGPYKCKAQVTSCSKYSRDFSFTIVDPVTSPVLNIMVIQTETDRHITLHCLSVNGSLP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title