Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allergin-1 Mouse anti-Mouse, Unconjugated, Clone: TX83, MilliporeSigma™

Catalog No. MABF23322MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23322MI 25 μL
MABF23321MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23322MI Supplier MilliporeSigma Supplier No. MABF233225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Mouse samples. It is validated for Flow Cytometry, Function Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Allergin-1
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone TX83
Conjugate Unconjugated
Gene Accession No. NP_001028607
Gene Alias Allergy inhibitory receptor 1;Mast cell antigen 32;MCA-32;Mast cell Ag-32;Mast cell immunoglobulin-like receptor 1
Gene Symbols Mirr1, Gm885, Mca32
Host Species Mouse
Immunogen BW5147 cells expressing Flag-tagged Allergin-1.
Molecular Weight of Antigen 27.37 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone TX83 is a mouse monoclonal antibody that detects murine Allergin-1.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.