Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Allergin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113085
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113085 Supplier Invitrogen™ Supplier No. PA5113085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Allergin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Allergin 1 is a protein coding gene. Immunoglobulin-like receptor which plays an inhibitory role in degranulation of mast cells. Negatively regulates IgE-mediated mast cell activation and suppresses the type I immediate hypersensitivity reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Allergin 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MILR1
Gene Accession No. Q3TB92, Q7Z6M3
Gene Alias Allergin-1; Allergy inhibitory receptor 1; C17orf60; Gm885; mast cell Ag-32; mast cell antigen 32; mast cell immunoglobulin like receptor 1; mast cell immunoglobulin-like receptor 1; MCA32; MCA-32; MCA32-1S2; Milr1; probable mast cell antigen 32 homolog; surface protein MCA-32
Gene Symbols MILR1
Host Species Rabbit
Immunogen Recombinant Human Allergin-1 protein (249-343AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284021, 380732
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.