Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allergin-1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987774
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7774 Supplier Bioss Supplier No. BS7348R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Allergin 1 is a protein coding gene. Immunoglobulin-like receptor which plays an inhibitory role in degranulation of mast cells. Negatively regulates IgE-mediated mast cell activation and suppresses the type I immediate hypersensitivity reaction.

Specifications

Antigen Allergin-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MILR1
Gene Accession No. Q7Z6M3
Gene Alias Allergin-1; Allergy inhibitory receptor 1; C17orf60; Mast cell antigen 32; mast cell immunoglobulin like receptor 1; Mast cell immunoglobulin-like receptor 1; MCA32; MCA-32; MCA32-1S2; MILR1; probable mast cell antigen 32 homolog
Gene Symbols MILR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Allergin-1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284021
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.