Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

allo-Inositol 98.0+%, TCI America™
SDP

Catalog No. I062925MG
Click to view available options
Quantity:
25 mg

Chemical Identifiers

CAS 643-10-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00799555
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
PubChem CID 892
ChEBI CHEBI:24848
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES C1(C(C(C(C(C1O)O)O)O)O)O

Specifications

Melting Point 320°C
Color White
Quantity 25 mg
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material allo-Inositol
EINECSNumber (3)-3265
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.