Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allophycocyanin Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8927V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8927V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8927V100 Supplier R&D Systems Supplier No. FAB8927V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Allophycocyanin Monoclonal specifically detects Allophycocyanin in Multi-Species samples. It is validated for Flow Cytometry.

Specifications

Antigen Allophycocyanin
Applications Flow Cytometry
Classification Monoclonal
Clone 936809
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P25054
Gene Alias APC
Host Species Mouse
Immunogen Allophycocyanin isolated from red algae Accession # P25054
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects Allophycocyanin in flow cytometry.
Target Species Multi-species
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.