Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allophycocyanin Antibody (936809), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310623 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310623 Supplier Novus Biologicals Supplier No. FAB8927APCCY7
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Allophycocyanin Monoclonal antibody specifically detects Allophycocyanin in Broad Species Reactivity samples. It is validated for Flow Cytometry

Specifications

Antigen Allophycocyanin
Applications Flow Cytometry
Classification Monoclonal
Clone 936809
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias APC
Host Species Mouse
Immunogen Allophycocyanin isolated from red algae, Accession # P25054
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.