Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Allopurinol, 98%

Catalog No. p-2695236
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 315-30-0 | C5H4N4O | 136.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315-30-0
Molecular Formula C5H4N4O
Molecular Weight (g/mol) 136.11
MDL Number MFCD00599413
InChI Key OFCNXPDARWKPPY-UHFFFAOYSA-N
Synonym allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
PubChem CID 2094
ChEBI CHEBI:40279
IUPAC Name 1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
SMILES O=C1N=CN=C2NNC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 384.0°C
Color White
Loss on Drying 0.5 % (1 g, 105°C) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 26,IV,2076
Merck Index 15,271
Solubility Information Solubility in water: 0.35g/L (25°C). Other solubilities: soluble in diluted alkalies and acids,practically insoluble in most common organic,solvents
Formula Weight 136.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allopurinol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or ras
EINECSNumber 206-250-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.