Learn More
Troemner™ Alloy 8 Metric Precision Weight Sets, Class 1
Catalog No. 02215125 Shop All Troemner Products
Click to view available options
Quantity:
12/Set
17/Set
21/Set
22/Set
23/Set
24/Set
Includes
Forceps
Excellent for high-precision calibrations and reference standards
Manufactured from a specially developed austenitic stainless-stee—Alloy 8—available exclusively from Troemner. Weights 1g and larger are manufactured using Alloy 8, which possesses closely controlled density, extremely low magnetic susceptibility, excellent stability and resistance to corrosion, assuring a very stable mass over time. Other weights are stainless-steel.
  • Each fractional weight is embossed with its value
  • Weights from 1 to 30mg are aluminum and flat, with one side turned up to form a handle for non-slip gripping with forceps
  • Weights from 50 to 500mg are stainless steel and flat with one side turned up to form a handle
  • Gram and kilogram weights are manufactured using Alloy 8 and come in cylindrical shapes with knobs on top for easy handling
  • Weight sets come in a 5-2-2-1 series, with two weights of any included mg or g value beginning with 2 (2mg, 20mg, etc.) and one weight for all other inclusive values
  • Certificate of calibration is available; contact your Fisher Customer Service Representative
TRUSTED_SUSTAINABILITY
Specifications
Product Type Medium Metric Analytical Weight Set
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 1 mg to 500 g
Mass 500 g, (2)200 g, 100 g, 50 g, (2)20 g, 10 g, 5g, (2)2 g, 1 g, 500 mg, (2)200 mg, 100 mg, 50 mg, (2)20 mg, 10 mg, 5 mg, (2)2 mg, 1 mg
Includes Stainless Steel cylindrical weight: 500g, 2 x 200g, 100g, 50g, 2 x 20g, 10g, 5g, 2 x 2g, 1g, Stainless Steel sheet weight: 500mg, 2 x 200mg, 100mg, 50mg, 2 x 20mg, 10mg, 5mg, Aluminum sheet weight: 2 x 2mg, 1mg, Polycarbonate case
Quantity 24/Set
Packaging Quantity Multiple
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.