Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allyl acetate, 97%
Click to view available options
Quantity:
50 mL
250 mL
Description
Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 6°C |
Density | 0.928 g/mL |
Boiling Point | 103°C to 104°C |
Refractive Index | 1.4042 |
Quantity | 250 mL |
UN Number | UN2333 |
Merck Index | 14,285 |
Solubility Information | Soluble in ethanol. Miscible with ethanol and ether. Slightly soluble in water. |
Formula Weight | 100.12 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H225-H301+H331-H312-H319 |
Hazard Statement | GHS H Statement H225-H301-H312 Highly flammable liquid and vapor. Toxic if swallowed. Harmful in contact with skin. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL ACETATE |
EINECSNumber | 209-734-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction