Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide

Catalog No. p-7081688
Click to view available options
Quantity:
250 g
1000 g

CAS: 106-95-6 | C3H5Br | 120.977 g/mol

Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

Solubility
Slightly miscible with water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-95-6
Molecular Formula C3H5Br
Molecular Weight (g/mol) 120.977
MDL Number MFCD00000244
InChI Key BHELZAPQIKSEDF-UHFFFAOYSA-N
Synonym allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide
PubChem CID 7841
IUPAC Name 3-bromoprop-1-ene
SMILES C=CCBr

Specifications

Melting Point °C to 119°C
Density 1.398
Boiling Point 70°C to 71°C
Flash Point -2°C (28°F)
Assay Percent Range 99%
Refractive Index 1.47
Linear Formula CH2=CHCH2Br
Quantity 250 g
UN Number UN1099
Sensitivity Light sensitive
Merck Index 14,288
Solubility Information Slightly miscible with water.
Formula Weight 120.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl Bromide, Stabilized with 300 to 1000ppm Propylene oxide
Show More Show Less
Hazard Category H225-H301+H331-H314-H335-H340-H350
Hazard Statement GHS H Statement
H225-H301-H331-H340-H350-H314
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: ALLYL BROMIDE
EINECSNumber 203-446-6
RTECSNumber UC7090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.