Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl chloride, 98%, stabilized

Catalog No. p-3598298
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 107-05-1 | C3H5Cl | 76.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-05-1, 75-56-9
Molecular Formula C3H5Cl
Molecular Weight (g/mol) 76.53
MDL Number MFCD00000984
InChI Key OSDWBNJEKMUWAV-UHFFFAOYSA-N
Synonym allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
PubChem CID 7850
ChEBI CHEBI:82379
IUPAC Name 3-chloroprop-1-ene
SMILES C=CCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -136.0°C
Color Colorless to Yellow
Density 0.9390g/mL
Boiling Point 44.0°C to 46.0°C
Flash Point −29°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2Cl
Refractive Index 1.4140 to 1.4160
Quantity 100 mL
Beilstein 01,198
Fieser 13,198
Merck Index 15,28
Specific Gravity 0.939
Solubility Information Solubility in water: 3.6g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol>50g/100mL,soluble in ch2cl2,acetone >50g/100 mL,soluble in n,n-dimethylformamide >50g/100 mL,miscible with chloroform and ether
Stabilizer 0.05 to 0.09% propylene oxide
Viscosity 0.34 mPa.s (20°C)
Formula Weight 76.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation. Suspected of causing cancer. Suspected of causing genetic defects. Harmful in contact with skin. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Harmful if inhaled. Very toxic to aquatic life. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge. Store in a well-ventilated place. Keep cool.
EINECSNumber 203-457-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.