Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allyl Cyanide 98%, Thermo Scientific™
Click to view available options
Quantity:
100 mL
500 mL
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Colorless to Yellow |
| Infrared Spectrum | Authentic |
| Linear Formula | H2C=CHCH2CN |
| Refractive Index | 1.4050 to 1.4070 (20°C, 589nm) |
| Quantity | 100 mL |
| Beilstein | 02, 408 |
| Merck Index | 15, 281 |
| Formula Weight | 67.09 |
| Show More |
Safety and Handling
| EINECSNumber | 203-701-1 |
| RTECSNumber | EM8050000 |
| TSCA | TSCA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction