Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Ethyl Ether 95.0+%, TCI America™
SDP

Catalog No. A103325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A103325ML 25 mL
A10335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A103325ML Supplier TCI America Supplier No. A103325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 557-31-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009255
InChI Key OJPSFJLSZZTSDF-UHFFFAOYSA-N
Synonym allyl ethyl ether, 1-propene, 3-ethoxy, allylethylether, ethyl allyl ether, unii-sm8t658re3, ether, allyl ethyl, 3-ethoxy-1-propene, ethyl 2-propenyl ether, 3-ethoxypropene, acmc-1atoi
PubChem CID 11191
IUPAC Name 3-ethoxyprop-1-ene
SMILES CCOCC=C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 25 mL
UN Number 2335
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Ethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.